Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-alpha-Phellandrene 65.0+%, TCI America™
SDP

Catalog No. M00515ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Catalog No. M00515ML

Supplier: TCI America M00515ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4221-98-1
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00062988
InChI Key OGLDWXZKYODSOB-SNVBAGLBSA-N
Synonym (-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene, (-)-p-Mentha-1,5-diene
PubChem CID 442482
ChEBI CHEBI:301
IUPAC Name (5R)-2-methyl-5-propan-2-ylcyclohexa-1,3-diene
SMILES CC1=CCC(C=C1)C(C)C

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 mL
UN Number 2319
Formula Weight 136.24
Percent Purity ≥65.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-alpha-Phellandrene
EINECSNumber (3)-2243
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.