Learn More
BEST SANITIZERS Alpet No-Rinse Quat Sanitizer, 1 gal
Supplier: BEST SANITIZERS SS10018
This item has a minimum qty of 3 per supplier requirements.
Alpet No-Rinse Quat Sanitizer is a dilutable sanitizer for food contact and non-food contact surfaces. With an NSF listed D2 classification, this product is an effective drain sanitizer and rubber footwear sanitizer. Dilutable, no-rinse surface sanitizer for food and non-food contact surfaces Broad spectrum, dual quat for dependable efficacy Sanitizes pre-cleaned food contact surfaces in 60 seconds and non-food contact surfaces in 30 seconds Disinfects in 10 minutes Effective footwear sanitizer Effective drain sanitizer NSF listed D2 classification EPA registered Kosher certified and Pareve
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.