Learn More
Abbott™ ID NOW Strep A 2 Test Kit
Shop All Abbott Rapid Diagnostics ProductsDescription
Abbott ID NOW Strep A 2 is now the fastest Strep A test without the need for culture confirmation. It provides positive results in as little as 2 minutes; allowing you to confidently test, and with a positive result, treat during the patient visit.
Faster than other molecular methods and more accurate than conventional rapid tests, Abbott ID NOW Strep A 2 delivers results within an actionable time frame allowing you to make real-time clinical decisions which impact patient care.
- Now CLIA-waived
- Offers exceptional performance
- Positive results in as little as 2 minutes
- Nearly 90% fewer false negatives than RADTs
- Enhanced molecular sensitivity
- Eliminates the need for culture confirmation of negative results
- Allows for the accurate detection of Group A Strep
- Increased detection compared to culture
- Eliminates interpretation and transcription errors
- Gives you the confidence to make clinical decisions sooner
- Facilitates effective patient management
- Small footprint enables ease of use in any healthcare setting
- Aids targeted antiviral therapy and Antimicrobial Stewardship
- Specimen type: throat swab
Specifications
Specifications
| Description | Strep A Test Kit |
| Sample Type | Throat Swab |
| Type | Strep A Test |
| For Use With (Application) | For accurate detection of Group A Strep |
| Quantity | 24 Test Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.