Learn More
Selleck Chemical LLC AG-1478 (Tyrphostin AG-1478) 5mg 153436-53-4 NSC 693255
Supplier: Selleck Chemical LLC S27285MG
AG-1478 (Tyrphostin AG-1478, NSC 693255) is a selective EGFR inhibitor with IC50 of 3 nM in cell-free assays, almost no activity on HER2-Neu, PDGFR, Trk, Bcr-Abl and InsR. AG-1478 (Tyrphostin AG-1478) inhibits encephalomyocarditis virus (EMCV) and hepatitis c virus (HCV) by targeting phosphatidylinositol 4-kinase III? (PI4KA). *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.