Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ AE150 Benchtop pH Meter

Catalog No. 13636ae153 Shop All Fisherbrand Products
Click to view available options
Type:
3-in-1 Set
Bio Set
Education Set
Standard Set

Catalog No. 13-636-AE153

Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price

Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet AE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.

  • One to five point pH calibration
  • Includes 13-620-AE6 3-in-1 single junction gel pH/ATC electrode
  • Automatic buffer recognition with two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Outputs Recorder (±2000 mV)
Accuracy (Temperature) ±0.3°C
Accuracy (mV) ±0.2 ±199.9 ±2 beyond ±200
Accuracy (pH) ±0.01
Includes Meter, 13-620-AE6 3-in-1 single-junction gel pH/ATC electrode, and stand
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Resolution (mV) 0.1 mV (±199.9 mV); 1 mV beyond ±200 mV
Calibration Points 1 to 5 with automatic buffer recognition (ABR)
Temperature Range (Metric) 0.0°C to 100.0°C
Resolution (Temperature) 0.1°C
Temperature Probe Calibration Offset in 0.1 °C increments; Offset range: ±5 °C
Memory 100 data sets
Inputs BNC, 2.5 mm phono plug, reference pin tip, ground
Warranty 3 Year Meter Warranty; 6 Month Electrode Warranty
Temperature Compensation Automatic or manual (0 to 100 °C)
Type 3-in-1 Set
Electrical Requirements AC/DC 9 V adapter (110/220 VAC, 50-60 Hz)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.