Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Advanced Instant Hand Sanitizer - Refreshing Gel
SDP

Catalog No. 19043961 Shop All Gojo Industries, Inc. Products
Click to view available options
Volume (English):
8 oz.
This item is not returnable. View return policy

Catalog No. 19-043-961

Supplier: PURELL™ 410212S

This item is not returnable. View return policy

PURELL™ Advanced Instant Hand Sanitizer - Refreshing Gel in a round bottle with pump head

  • PURELL™ Advanced Instant Hand Sanitizer - Refreshing Gel
  • Kills 99.99% of most common germs that may cause illness
  • Outperforms other hand sanitizers ounce for ounce
  • Clinically proven to maintain skin health
  • America’s #1 Instant Hand Sanitizer
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Form Gel
Quantity 12/Cs.
Volume (English) 8 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.