Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADH1A/ADH1B/ADH1C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116865
Description
Positive Samples: Mouse lung Immunogen sequence: IGRLDTMMAS LLCCHEACGT SVIVGVPPAS QNLSINPMLL LTGRTWKGAV YGGFKSKEGI PKLVADFMAK KFSLDALITH VLPFEKINEG FDLLHSGKSI R.
Alcohol dehydrogenase,consisting of several homo- and heterodimers of alpha, beta, and gamma subunits, exhibits high activity for ethanol oxidation and plays a major role in ethanol catabolism.Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Three genes encoding alpha, beta and gamma subunits are tandemly organized in a genomic segment as a gene cluster.
Specifications
| ADH1A/ADH1B/ADH1C | |
| Polyclonal | |
| Unconjugated | |
| ADH1C | |
| ADH, alpha subunit; ADH, beta subunit; ADH, gamma subunit; ADH1; ADH1A; ADH1B; ADH1C; ADH2; ADH3; alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 1B; alcohol dehydrogenase 1B (class I), beta polypeptide; alcohol dehydrogenase 1C; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase 3 (class I), gamma polypeptide; alcohol dehydrogenase subunit alpha; Alcohol dehydrogenase subunit beta; Alcohol dehydrogenase subunit gamma; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117; HEL-S-117 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 124, 125, 126 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.297 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P00325, P00326, P07327 | |
| ADH1A, ADH1B, ADH1C | |
| A synthetic peptide corresponding to a sequence within amino acids 270-370 of human ADH1A/ADH1B/ADH1C (NP_0006592). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction