Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Adflo™ PAPR and Versaflo™ M-Series Helmet Kit
Shop All 3M Products
Click to view available options
Includes:
No ADF
Description
- Able to lift visors in non-hazardous areas to help facilitate communicationsProtection from welding arc (ANSI Z87) plus spark and splatter
- Head top can be used for most applications including arc gouging, plasma cutting, grinding, MIG, TIG, and GMAW weldingProtection from welding arc (ANSI Z87) plus spark and splatter
- When used in conjunction with NIOSH approved air source, is designed to help protect the user from certain airborne contaminants, and certain head, eye, face, and welding hazardsProtection from welding arc (ANSI Z87) plus spark and splatter
- Compatible with 9100XXi ADF with Natural Color Technology
- 3M Catalog# 38-1101-00SW
Specifications
Specifications
| Certifications/Compliance | ANSI Z89.1 Type I, Class G |
| Color | Black |
| For Use With (Application) | Welding |
| Includes | No ADF |
| Material | Leather |
| Model | 38-1101-00SW |
| Product Type | Welding Respiratory System |
| Quantity | 1 Ea. |
| Size | Standard |
| Visor Type | Clear |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction