Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine 5'-Triphosphate, [γ-32P]Ultratide, MP Biomedicals
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
250 μCi
Description
Recommended Storage
Store at -20°C
Specifications
Specifications
| Quantity | 250 μCi |
| Formula Weight | 912 |
| Chemical Name or Material | Adenosine 5'-Triphosphate, [γ-32P]Ultratide, Tetra-triethylammonium Salt |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction