Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 97-120 Oil Impermeable Industrial Gloves

Catalog No. 19501053 Shop All Ansell Products
Click to view available options
Size:
8
9
10
11
12

Catalog No. 19-501-053

Supplier: Ansell 97120080

ActivArmr™ gloves protect hands in extreme applications with specialized oil impermeable technology, and impact resistant construction.

  • Oil Impermeable Barrier repels drilling fluids and oils to keep hands clean and dry
  • Mud-oil grip with cut and impact resistance protects hands, reduces fatigue, and increases productivity
  • Hi-vis gloves feature a reflective logo for enhanced visibility
  • Impact bumpers protect entire back of hand from medium-level impact injury
  • Ideal for rugged environments and extreme applications
  • All-day comfort that helps workers get the job done
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Back Material PVC
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A2
Hand Specific
Material Nylon, Spandex
Product Type Mechanic's Gloves
Size 8
EN388 Standard 4242BP
For Use With (Application) Oil and Gas Drilling
Glove Cuff Style Knit
Liner Material Aramid Fiber
Packaging Type Polybag
Series 97-120
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.