Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Violet 43 97.0+%, TCI America™
SDP

Catalog No. A140225G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Catalog No. A140225G

Supplier: TCI America A140225G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4430-18-6
Molecular Formula C21H14NNaO6S
Molecular Weight (g/mol) 431.394
MDL Number MFCD00068446
InChI Key GTKIEPUIFBBXJQ-UHFFFAOYSA-M
PubChem CID 23669622
IUPAC Name sodium;2-[(4-hydroxy-9,10-dioxoanthracen-1-yl)amino]-5-methylbenzenesulfonate
SMILES CC1=CC(=C(C=C1)NC2=C3C(=C(C=C2)O)C(=O)C4=CC=CC=C4C3=O)S(=O)(=O)[O-].[Na+]

Specifications

Formula Weight 431.39
Percent Purity ≥97.0% (HPLC)
Color Purple
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Acid Violet 43
EINECSNumber (5)-1608
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.