Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, Technical, Reagents
Click to view available options
Quantity:
4 L
20 L
55 gal.
Specifications
Specifications
| Melting Point | -94.7°C |
| Color | Colorless |
| Boiling Point | 56°C |
| Quantity | 20 L |
| Packaging | Steel Pail |
| Physical Form | Liquid |
| Specific Gravity | 0.788 |
| Grade | Technical |
| Chemical Name or Material | Acetone, Technical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction