Learn More
Electron Microscopy Sciences Acetone, Glass Distilled 100 ML
SDP
Supplier:  Electron Microscopy Sciences 10015
Encompass_Preferred
(2-Propanone , DMK , Dimethyl ketone , Dimethylformaldehyde , Propanone)
CAS #67-64-1 CH₃COCH₃ F.W. 58.08 g/mol
U.V. Absorption (1 cm cell vs. H2O) is 0.01, 0.02, 0.10, and 1.0 at wavelengths of 400, 350, 340, and 330.
Catalog No. 50-980-142