Learn More
Electron Microscopy Sciences Acetate Film 35 µm, 10x15 cm sheet
Supplier: Electron Microscopy Sciences 5042030
This acetate film soften in acetone and can be used for duplicating purposes. Acetate film which is wet with acetone can then be applied to a surface which has been wet with acetone. It is easily removed away from the object when dry. Acetate films come in 35um thickness. Softens in acetone, can be used for duplicating purposes. Wet film with acetone then be apply to surface which has been wet with acetone, easily removed from the object when dry. Comes in 35 micron thickness.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.