Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Glutathione Assay Kit

Catalog No. 89028381 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit

Catalog No. 89-028-381

Supplier: Abnova™ KA1649

Only null left
Add to Cart
Add to Cart

Colorimetric determination of reduced glutathione at 412nm in blood, cell, plasma, serum, tissue, urine

Specifications

Calibration Range 0.4 to 100μM
Content And Storage Store the kit at 4°C.
Description Glutathione Assay Kit is a colorimetric determination of reduced glutathione at 412 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 30mL Reagent A, 30mL reagent B, 10mL 100μM Glutathione calibrator
Quantity 1 Kit
Sample Type Blood, Cell, Plasma, Serum, Tissue, Urine
Sample Volume 120 μL
Type Detection Assay Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.