Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
GSA_VAGREENER_CHOICE
Catalog No. 1424571
Click to view available options
Material:
Polystyrene
Vinyl
Well Shape:
Conical Bottom (V-well)
Curved Edges, Flat Bottom (C-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Volume (Metric) Well:
280 μL
330 μL
400 μL
Lid:
With Lid
Without Lid
This item is not returnable. View return policy
Catalog No. 14-245-71
Supplier: Thermo Scientific™ 2205
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Features:
  • Use for high-throughput screening and research
  • Clear plates for colorimetric applications
  • Standard footprint, 128 × 86mm
  • Available as both polystyrene square wells and vinyl square wells
  • Vinyl Microtiter microplates are used for complement fixation, agglutination and blood typing
  • Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Hydrophobic
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 50 Cases, 10 Pk.
Product Line Microtiter
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 280 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
No. per Case 50
No. per Pack 10
Quantity 50/Pk.
Well Design U96
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.