Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Decyn-1-ol 94.0+%, TCI America™
SDP

Catalog No. D37105G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. D37105G

Supplier: TCI America D37105G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17643-36-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00049199
InChI Key KOAOUMQUUZNRLL-UHFFFAOYSA-N
Synonym 10-Hydroxy-1-decyne
PubChem CID 543752
IUPAC Name dec-9-yn-1-ol
SMILES C#CCCCCCCCCO

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 154.25
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 9-Decyn-1-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.