Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
9-Bromofluorene, 97+%
Click to view available options
Quantity:
5 g
50 g
Description
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
Specifications
Specifications
| Melting Point | 101°C to 105°C |
| Odor | Irritating |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 2047220 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 245.12 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 9-Bromofluorene |
Safety and Handling
| Hazard Category | H314-H317 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 217-722-9 |
| RTECSNumber | LL5890000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction