Learn More
Chemglass Life Sciences 80mm Envelope Style Gasket
Supplier: Chemglass Life Sciences CG147G02
80mm Envelope Style Gasket envelope style gaskets are machined using chemically resistant virgin PTFE and have a silicone core. The larger sealing surface results in improved sealing under full vacuum, as opposed to other o-ring materials. The machined in o-ring aids in sealing and centering of gasket on the Schott o-ring flange.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.