Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Undecenal, predominantly cis, 96%, Thermo Scientific™

Catalog No. AAA1756106
Click to view available options
Quantity:
5 g
25 g
100 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58296-81-4
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00015577
InChI Key DWQXOILNGUCNSH-ARJAWSKDSA-N
Synonym cis-8-undecen-1-al, 8-undecenal, 8z, 8-undecenal, unii-u4oq6978qw, z-8-undecenal, 8-undecen-1-al high cis, cis-8-undecenal, z-undec-8-enal, 8z-undec-8-enal
PubChem CID 6436710
IUPAC Name (Z)-undec-8-enal
SMILES CCC=CCCCCCCC=O

Specifications

Melting Point 261°C to 263°C
Density 0.845
Boiling Point 95°C (4 mmHg)
Flash Point 93°C (199°F)
Quantity 5 g
Formula Weight 168.29
Percent Purity 96%
Chemical Name or Material 8-Undecenal, predominantly cis
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 261-202-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.