Learn More
Electron Microscopy Sciences 8% Paraformaldehyde (formaldehyde) aqueous solution, 100mL
Supplier: Electron Microscopy Sciences 1578100
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.