Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 8-CPT-2Me-cAMP, sodium salt
GREENER_CHOICE

Catalog No. 16451
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy

Catalog No. 16-451

Supplier: Tocris Bioscience™ 1645/1

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Epac activator

Selective activator of Epac, the cAMP-sensitive guanine nucleotide-exchange factor for Rap1 and Rap2. Activates Epac1 (EC50 = 2.2 μM), but not PKA (EC50 > 10 μM). Stimulates Epac-mediated Ca2+-mediated Ca2+ release in pancreatic β-cells in vitro. Cell permeable analog (Cat. No. 4853) also available.

Chemical Identifiers

CAS 634207-53-7
Molecular Formula C17H17ClN5NaO6PS

Specifications

Quantity 1 mg
Formula Weight 507.82
Purity >99%
Molecular Weight (g/mol) 508.826
Chemical Name or Material EPAC Activators
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.