Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega 6 x 5 LC-MS/MS Peptide Reference Mix

Catalog No. p-7068012 Shop All Promega Corporation Products
Click to view available options
Quantity:
200 pmol
50 μL

Catalog No. PRV7495

Supplier: Promega V7495

Only null left
Add to Cart
Add to Cart

The 6 x 5 LC-MS/MS Peptide Reference Mix is a unique reagent designed to monitor liquid chromatography (LC) and mass spectrometry (MS) instrument performance and assist in method development and optimization.

  • Peptide Mix and Free Software to Monitor LC/MS Instrument Performance
  • Determine mass spec instrument sensitivity and dynamic range
  • No manual calculations, maximized instrument performance with free PReMiS™ Software

Specifications

Content And Storage -30°C to -10°C
Quantity 200 pmol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.