Learn More
Hach Company Sension+ 5060 portable platinum conductivity cell for general purpose applications
Supplier: Hach Company LZW5060.97.0002
Sension+ 5060 is a three-pole platinum conductivity cell with a polycarbonate body and built-in temperature sensor. The 5060 has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable Conductivity meters. The 5060 is ideal for conductivity measurements in general aqueous applications.