Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-1-hexyne, 98%

Catalog No. AAL0601406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0601406 5 g
AAL0601414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0601406 Supplier Thermo Scientific Chemicals Supplier No. L0601406
Only null left
Add to Cart

CAS: 2203-80-7 | C7H12 | 96.173 g/mol

5-Methyl-1-hexyne is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1-hexyne is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2203-80-7
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00041610
InChI Key HKNANEMUCJGPMS-UHFFFAOYSA-N
Synonym 5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa
PubChem CID 137490
IUPAC Name 5-methylhex-1-yne
SMILES CC(C)CCC#C

Specifications

Density 0.732
Boiling Point 91°C to 93°C
Flash Point −4°C (24°F)
Refractive Index 1.4075
Quantity 5 g
UN Number UN3295
Beilstein 1731845
Solubility Information Not miscible in water.
Formula Weight 96.17
Percent Purity 98%
Chemical Name or Material 5-Methyl-1-hexyne
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.