Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione 98.0+%, TCI America™
SDP

Catalog No. h11631g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Catalog No. H11631G

Supplier: TCI America H11631G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 202126-51-0
Molecular Formula C10H14S5
Molecular Weight (g/mol) 294.52
MDL Number MFCD09038526
InChI Key RMWMTOWGAZGYGL-UHFFFAOYSA-N
PubChem CID 44629827
IUPAC Name 5-hexyl-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2-thione
SMILES CCCCCCC1SC2=C(S1)SC(=S)S2

Specifications

Quantity 1 g
Formula Weight 294.52
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hexyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.