Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylstyrene, 94%, stab. with 50 ppm 4-tert-butylcatechol

Catalog No. AAL12356AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL12356AE 100 mL
AAL12356AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL12356AE Supplier Thermo Scientific Chemicals Supplier No. L12356AE
Only null left
Add to Cart

CAS: 1746-23-2 | C12H16 | 160.26 g/mol

4-tert-Butylstyrene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylstyrene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in the dark. Protect from heat. Incompatible with heat, light, free radical initiators, UV radiation. Do not store together.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1746-23-2
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00065126
InChI Key QEDJMOONZLUIMC-UHFFFAOYSA-N
Synonym 4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
PubChem CID 15627
IUPAC Name 1-tert-butyl-4-ethenylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C=C

Specifications

Melting Point -38°C
Density 0.884
Boiling Point 219°C
Flash Point 80°C (176°F)
Refractive Index 1.525
Quantity 100 mL
Beilstein 1903714
Solubility Information Not miscible in water.
Formula Weight 160.26
Percent Purity 94%
Chemical Name or Material 4-tert-Butylstyrene, stabilized with 50ppm 4-tert-butylcatechol
Show More Show Less
Hazard Category H227-H315-H319-H332
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 217-126-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.