Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Nitrophenetole 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
Specifications
Specifications
| Melting Point | 59°C |
| Color | Yellow |
| Boiling Point | 112°C |
| Quantity | 25 g |
| Formula Weight | 167.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrophenetole |
Safety and Handling
| EINECSNumber | (3)-0783 |
| RTECSNumber | DA0600000 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction