Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methyl-1-cyclohexene, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
500 mL
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 104°C |
| Quantity | 25 mL |
| UN Number | 3295 |
| Formula Weight | 96.17 |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Methyl-1-cyclohexene |
Safety and Handling
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction