Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzenesulfonic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. C060625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Catalog No. C060625G

Supplier: TCI America C060625G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98-66-8
Molecular Formula C6H5ClO3S
Molecular Weight (g/mol) 192.613
MDL Number MFCD00065342
InChI Key RJWBTWIBUIGANW-UHFFFAOYSA-N
Synonym p-chlorobenzenesulfonic acid, 4-chloro-benzenesulfonic acid, benzenesulfonic acid, 4-chloro, p-chlorophenylsulfonic acid, unii-5cn6b87ik2, 4-chlorobenzenesulphonic acid, benzenesulfonic acid, p-chloro, 4-chlorobenzene sulphonic acid, 4-chlorobenzene sulfonic acid, closylate
PubChem CID 7400
IUPAC Name 4-chlorobenzenesulfonic acid
SMILES C1=CC(=CC=C1S(=O)(=O)O)Cl

Specifications

Quantity 25 g
UN Number 2585
Formula Weight 192.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzenesulfonic Acid Hydrate
RTECSNumber DB5074000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.