Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dibromo-2-(6-bromohexyl)benzotriazole 97.0+%, TCI America™
SDP

Catalog No. D4514200MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg

Catalog No. D4514200MG

Supplier: TCI America D4514200MG

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 890704-02-6
Molecular Formula C12H14Br3N3
Molecular Weight (g/mol) 439.977
MDL Number MFCD19440927
InChI Key VWPRIVLLHDPAJM-UHFFFAOYSA-N
PubChem CID 71721496
IUPAC Name 4,7-dibromo-2-(6-bromohexyl)benzotriazole
SMILES C1=C(C2=NN(N=C2C(=C1)Br)CCCCCCBr)Br

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 439.98
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-2-(6-bromohexyl)benzotriazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.