Learn More
4,4'-Dibromobenzophenone, 98+%
Description
4,4'-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,4′-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.
Solubility
Soluble in ethanol, 2-propanol, acetone and toluene. Miscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | 172°C to 176°C |
| Molecular Formula | C13H8Br2O |
| MDL Number | MFCD00016330 |
| Quantity | 1 g |
| Beilstein | 2049958 |
| Synonym | 4,4'-dibromobenzophenone, bis 4-bromophenyl methanone, methanone, bis 4-bromophenyl, p,p'-dibromobenzophenone, bis p-bromophenyl ketone, 4,4-dibromobenzophenone, di4-bromophenyl ketone, bis-4-bromo-phenyl-methanone, zlchem 265, pubchem7494 |
| Solubility Information | Soluble in ethanol,2-propanol,acetone and toluene. Miscible with water. |
| InChI Key | LFABNOYDEODDFX-UHFFFAOYSA-N |
| SMILES | BrC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1 |
| IUPAC Name | bis(4-bromophenyl)methanone |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 223-632-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.