Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Chemical Sorbent Mini-Boom P-212

Catalog No. 19039443 Shop All 3M Products
Change view
Click to view available options
Capacity (English):
3 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-039443 3 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Failure to properly handle and dispose of sorbents contaminated by hazardous materials can result in serious physical injury or death and/or environmental harm. For proper handling, consult the Material Safety Data Sheet for the material absorbed. To reduce the risk of serious physical injury or death, always use respiratory protection and protective clothing appropriate for handling the material to be absorbed.
Catalog No. 19-039443 Supplier 3M™ Supplier No. 7000001921
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Failure to properly handle and dispose of sorbents contaminated by hazardous materials can result in serious physical injury or death and/or environmental harm. For proper handling, consult the Material Safety Data Sheet for the material absorbed. To reduce the risk of serious physical injury or death, always use respiratory protection and protective clothing appropriate for handling the material to be absorbed.

Used to encircle spills and to contain leaks

Chemical Sorbent Mini-Booms can be placed around hazardous spills, reducing the possibility of spread to adjacent areas. Chemical Sorbents can sorb acids, bases and other hazardous fluids. Three gallon capacity.

  • For use on most aggressive (hazardous) and nonaggressive liquids
  • Ideal for placing around equipment
  • Helps prevent leaks/spills from spreading to adjacent areas
  • Scrim on the outside of mini-boom identifies the use while containing particulate Sorbent material
  • Measures: D x L: 3 in. x 12 ft. (7.6cm x 3.6m)
  • 3M Catalog# P212

Applications:

Agriculture, Automotive, Chemicals, Construction, General Manufacturing, Healthcare, Military, Mining, Oil & Gas, Pharmaceuticals, Primary Metals, Transportation Industries

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 3 gal.
Capacity (Metric) 11.35 L
Type Chemical Sorbent Mini Boom
For Use With (Application) Absorbs acids, bases and other hazardous fluids
Model P212
Length (English) 12 ft.
Length (Metric) 3.65 m
Diameter (English) 3 in.
Diameter (Metric) 7.62 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.