Learn More
MSC Loctite 680522 0.14 oz Bottle Clear Instant Adhesive Series QuickTite, 5 to 30 sec Working Time, 24 hr Full Cure Time, Bonds to Metal & Plastic
Supplier: MSC 08844797
Loctite 680522 0.14 oz Bottle Clear Instant Adhesive Series QuickTite, 5 to 30 sec Working Time, 24 hr Full Cure Time, Bonds to Metal & Plastic
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.