Learn More
Ace Glass, Inc. Gauge, pressure, complete, 0-400psi, stainless steel internal materials, 1/4in male NPT bottom fitting, 2.5in diameter. Comes with one 13385-20 nylon adapter and one 12720-25, 3-way, PTFE tubing connector.
Supplier: Ace Glass, Inc. 1338538
0-400psi pressure gauge with stainless steel internal materials. Features male NPT bottom fitting. Gauges diameter is 2.5 inches. Gauge comes with one nylon adapter (ACE 13385-20) and one 3-way PTFE tubing connector (ACE 12720-25).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.