Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Decylthiophene 98.0+%, TCI America™
SDP

Catalog No. D20981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20981G 1 g
D20985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20981G Supplier TCI America Supplier No. D20981G

Chemical Identifiers

CAS 65016-55-9
Molecular Formula C14H24S
Molecular Weight (g/mol) 224.406
MDL Number MFCD00143180
InChI Key JAYBIBLZTQMCAY-UHFFFAOYSA-N
Synonym 3-n-decylthiophene, 3-decylthiophene, thiophene, 3-decyl, acmc-1bell, ksc491i4d
PubChem CID 566842
IUPAC Name 3-decylthiophene
SMILES CCCCCCCCCCC1=CSC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 224.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Decylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.