Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropivalic Acid 98.0+%, TCI America™
SDP

Catalog No. C155425G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Catalog No. C155425G

Supplier: TCI America C155425G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13511-38-1
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00004195
InChI Key YBJGQSNSAWZZHL-UHFFFAOYSA-N
Synonym 3-Chloro-2,2-dimethylpropionic Acid
PubChem CID 83542
IUPAC Name 3-chloro-2,2-dimethylpropanoic acid
SMILES CC(C)(CCl)C(O)=O

Specifications

Melting Point 43°C
Color White
Boiling Point 131°C
Quantity 25 g
UN Number 3261
Formula Weight 136.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloropivalic Acid
EINECSNumber (2)-1180
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.