Learn More
YUKON MEDICAL 20mm Vialok Vented with Large Vent (CS/100)
Supplier: YUKON MEDICAL YM020
20mm Vialok Vented Vial Access Devices feature a 0.2 micron filter that neutralizes vial pressure and minimizes aerosols and surface contamination. In addition to their superior vial security, they utilize a dual lumen spike, making for efficient transfers and minimizing the drug residual left in the vial.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.