Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiouracil, 99%, Thermo Scientific Chemicals

Catalog No. p-3758824
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle

Catalog No. AC151840050

Supplier: Thermo Scientific Chemicals 151840050

Only null left
Add to Cart

2-Thiouracil, Thiouracil or 4-Hydroxy-2-mercaptopyrimidine, CAS # 141-90-2, is the modified analog of the nucleotide base, uracil.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description


• 2-Thiouracil (Thiouracil or 4-Hydroxy-2-mercaptopyrimidine) is a nucleotide analog with a C-2 thioxo substitution
• 2-Thiouracil is a melanocyte suppressor and thyroid inhibitor


Applications
• 2-Thiouracil can be used to inhibit nitric oxide synthase and suppresses nitric oxide signaling in vitro
• 2-Thiouracil can be used for labeling of RNA in vitro
• 2-Thiouracil is intended for research applications only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-90-2
Molecular Formula C4H4N2OS
Molecular Weight (g/mol) 128.15
MDL Number MFCD00006039
InChI Key ZEMGGZBWXRYJHK-UHFFFAOYSA-N
Synonym 2-thiouracil, thiouracil, 4-hydroxy-2-mercaptopyrimidine, antagothyroid, antagothyroil, deracil, nobilen, 2-thioxo-2,3-dihydropyrimidin-4 1h-one, 2-mercaptopyrimidin-4-ol, 2-mercapto-4-hydroxypyrimidine
PubChem CID 1269845
ChEBI CHEBI:348530
IUPAC Name 2-sulfanylidene-1H-pyrimidin-4-one
SMILES O=C1NC(=S)NC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White to Cream
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 323
Merck Index 15, 9520
Solubility Information Solubility in water: 0.5g/L. Other solubilities: soluble in alkaline solutions
Formula Weight 128.15
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Thiouracil
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-508-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.