Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-iodobenzoic acid, 97%

Catalog No. aah2617503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2617503 1 g
AAH2617506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2617503 Supplier Thermo Scientific Chemicals Supplier No. H2617503
Only null left
Add to Cart

CAS: 124700-41-0 | C7H4FIO2 | 266.01 g/mol

2-Fluoro-5-iodobenzoic acid is one of the precursor molecules were shown to generate ETD reagents via ESI followed by CID. Starting material in the synthesis of 3, 6-disubstituted 2-pyridinecarboxamide(GK activators).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-iodobenzoic acid is one of the precursor molecules were shown to generate ETD reagents via ESI followed by CID. Starting material in the synthesis of 3, 6-disubstituted 2-pyridinecarboxamide(GK activators).

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124700-41-0
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD03094517
InChI Key QNNJHBNTHVHALE-UHFFFAOYSA-N
Synonym benzoic acid, 2-fluoro-5-iodo, 2-fluoro-5-iodobenzoicacid, 2-fluoro-5-iodo-benzoic acid, pubchem1385, acmc-1c24w, ksc174o6d, rarechem al bo 1479, buttpark 122\01-06, 5-iodo-2-fluorobenzoic acid, 2-fluoro-5-iodobenzoic acid
PubChem CID 2778925
IUPAC Name 2-fluoro-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)F

Specifications

Melting Point 162°C to 165°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.