Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-[3-(2,4-dichlorophenyl)isoxazol-5-yl]ethan-1-one, 95%, Thermo Scientific™

Catalog No. SPB04881EA
Change view
Click to view available options
:
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SPB04881EA Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB04881EA Supplier Thermo Scientific Chemicals Supplier No. SPB04881EA

Specifications

CAS 175334-69-7
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C11H6BrCl2NO2
Quantity 10g
MDL Number MFCD00173932
Synonym 2-bromo-1-3-2,4-dichlorophenyl isoxazol-5-yl ethanone, 2-bromo-1-3-2,4-dichlorophenyl isoxazol-5-yl ethan-1-one, 2-bromo-1-3-2,4-dichlorophenyl-1,2-oxazol-5-yl ethanone, 5-bromoacetyl-3-2,4-dichlorophenyl isoxazole, 2-bromo-1-3-2,4-dichlorophenyl-1,2-oxazol-5-yl, pubchem23339, buttpark 97\12-38, 1-3-2,4-dichlorophenyl isoxazol-5-yl-2-bromoethan-1-one
InChI Key WYJZTLHKOJBVSA-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)C2=NOC(=C2)C(=O)CBr
IUPAC Name 2-bromo-1-[3-(2,4-dichlorophenyl)-1,2-oxazol-5-yl]ethanone
Molecular Weight (g/mol) 334.978
PubChem CID 2799576
Formula Weight 334.98
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-[3-(2, 4-dichlorophenyl)isoxazol-5-yl]ethan-1-one
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.