Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluoropyridine 98.0+%, TCI America™
SDP

Catalog No. D41741G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. D41741G

Supplier: TCI America D41741G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84476-99-3
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD04114191
InChI Key XFAMUOYNXFXQTC-UHFFFAOYSA-N
PubChem CID 2762871
IUPAC Name 2,5-difluoropyridine
SMILES C1=CC(=NC=C1F)F

Specifications

Melting Point -33°C
Color Yellow
Boiling Point 115°C
Quantity 1 g
UN Number 1993
Synonym pyridine, 2,5-difluoro, 2,5-difluorpyridin, pubchem14398, acmc-209put, pyridine,2,5-difluoro, 2,5-bis fluoranyl pyridine, ksc652g9r, 2,5-difluoropyridine, abbypharma ap-14-5383
Formula Weight 115.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluoropyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.