Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-dimethylamino-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D22361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22361G 1 g
D22365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22361G Supplier TCI America Supplier No. D22361G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1985-46-2
Molecular Formula C5H10N6
Molecular Weight (g/mol) 154.177
MDL Number MFCD00191338
InChI Key IEFWDQQGFDLKFK-UHFFFAOYSA-N
Synonym N2,N2-Dimethylmelamine
PubChem CID 16134
IUPAC Name 2-N,2-N-dimethyl-1,3,5-triazine-2,4,6-triamine
SMILES CN(C)C1=NC(=NC(=N1)N)N

Specifications

Color White
Quantity 1 g
Formula Weight 154.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-dimethylamino-1,3,5-triazine
RTECSNumber OS0875000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.