Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorostyrene, 98%, stab. with 250ppm 4-tert-butylcatechol

Catalog No. AAA1683306
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAA1683306

Supplier: Thermo Scientific Chemicals A1683306

Only null left
Add to Cart

CAS: 653-34-9 | C8H3F5 | 194.104 g/mol

2,3,4,5,6-Pentafluorostyrene is used in the synthesis of optically active copolymers having low surface energies by utilizing beta-pinene as another monomer. It is also used in the synthesis of fluorinated and photo crosslinkable liquid prepolymers, which is used for flexible optical waveguides. Polymeric fluorinated nano particles prepared using 2,3,4,5,6-Pentafluorostyrene and styrene is explored in magnetic resonance image (MRI) applications, since it has larger structural design potential compared to traditional systems like emulsions and solutions of smaller molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorostyrene is used in the synthesis of optically active copolymers having low surface energies by utilizing beta-pinene as another monomer. It is also used in the synthesis of fluorinated and photo crosslinkable liquid prepolymers, which is used for flexible optical waveguides. Polymeric fluorinated nano particles prepared using 2,3,4,5,6-Pentafluorostyrene and styrene is explored in magnetic resonance image (MRI) applications, since it has larger structural design potential compared to traditional systems like emulsions and solutions of smaller molecules.

Solubility
Immiscible with water.

Notes
Store in a cool and well ventilated area. Keep the container tightly closed, away from sources of ignition and direct sunlight. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-34-9
Molecular Formula C8H3F5
Molecular Weight (g/mol) 194.104
MDL Number MFCD00000300
InChI Key LVJZCPNIJXVIAT-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
PubChem CID 69556
IUPAC Name 1-ethenyl-2,3,4,5,6-pentafluorobenzene
SMILES C=CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Density 1.406
Boiling Point 139°C to 140°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.446
Quantity 5 g
UN Number UN1993
Beilstein 1874390
Solubility Information Immiscible with water.
Formula Weight 194.1
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorostyrene, stabilized with 250ppm 4-tert-butylcatechol
Show More Show Less
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 211-500-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.