Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromoethyl)-2-methyl-1,3-dioxolane 96.0+%, TCI America™
SDP

Catalog No. B15985G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. B15985G

Supplier: TCI America B15985G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37865-96-6
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00191321
InChI Key SZLPRRVCSGZARF-UHFFFAOYSA-N
PubChem CID 11805619
IUPAC Name 2-(2-bromoethyl)-2-methyl-1,3-dioxolane
SMILES CC1(OCCO1)CCBr

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
Formula Weight 195.06
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromoethyl)-2-methyl-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.