Learn More
Newcomer Supply ACTIVATE, 10:1 Bleach Sprayer. Automatically meters and dispenses a 10% bleach solution with each trigger pull
SDP
Supplier:  Newcomer Supply 5101
Encompass
Bleach Sprayer, with refillable 11oz. Water cartridge. Bleach cartridge not included. Dual-liquid sprayer automatically combines water and bleach to make fresh 10% solution when you use it! Prevents bleach from premature degradation. Bleach and water are in separate containers until the trigger is squeezed. Refillable water cartridge & replaceable bleach cartridge. The Activate™ Bleach Sprayer Dilution System has been made to meter and dispense 5.25% Sodium Hypochlorite NaOCL at a 10% (9:1) solution. It has been widely recommended by CDC and other nationally recognized authorities.
Catalog No. NC1094882