Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1000 μg/mL Cannabidiol (CBD) Standard in P&T Methanol Solvent, Restek
SDP

Catalog No. 06722452 Shop All Restek Corporation Products
Click to view available options
Quantity:
1 mL

Catalog No. 06-722-452

Supplier: Restek Corporation 34011

Only null left
Add to Cart
Add to Cart
  • U.S. DEA-exempted formulation
    • No additional customer permits/licensing required to purchase within the U.S.
  • Canadian test kit registration numbers are assigned and available on individual product pages

Specifications

CAS 13956-29-1
Shipping Condition On ice
Concentration 1000 μg/mL
Certifications/Compliance Certified reference material
Type Cannabinoids Standard
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.