Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP30L Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277931
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7931 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7931 Supplier Leinco Technologies Inc Supplier No. 28113100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SAP30L Polyclonal antibody specifically detects SAP30L in Human samples. It is validated for Western Blot

Specifications

Antigen SAP30L
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SAP30L
Gene Accession No. Q9HAJ7
Gene Alias 2310079P12Rik; AF006998; DKFZp667L2214; FLJ11526; FLJ23595; FLJ36497; HCV non-structural protein 4A-transactivated protein 2; Histone deacetylase complex subunit SAP30L; L55; NS4ATP2; SAP30 like; SAP30L; SAP30-like; Sin3 corepressor complex subunit SAP30L; Sin3A associated protein p30-like; sin3A-associated protein p30-like protein; sin3-associated protein p30-like
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 165-183 (VKSNKSRLDQKSEGGKQLE) of human SAP30L conjugated to KLH at the N-terminus.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79685
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.