Learn More
Paper Thermometer Co 1-Temp THERMOLABEL 180°F/82°C Wide
Supplier: Paper Thermometer Co TL1180W
This item has a minimum order quantity of 12 packs per supplier requirements.
The 1-Temp Thermolabel Wide-180°F/82°C is a waterproof indicator with extra space on each label for the test date and tester's initials. Commonly used in research facilities, the temperatures sensitive square changes from white to black at 180°F. The permanent color change is now a record that the temperature has been reached. Store in a cool, dry place.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.