Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1H-indole-7-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC66904CB
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Catalog No. CC66904CB

Supplier: Thermo Scientific Chemicals CC66904CB

Only null left
Add to Cart

Chemical Identifiers

CAS 69047-36-5
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD11109329
InChI Key JOBIAZYEJOMXEP-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-7-carbaldehyde, 1h-indole-7-carboxaldehyde, 1-methyl, 1-methyl-7-formylindole, n-methylindole-7-carboxaldehyde, jobiazyejomxep-uhfffaoysa, 1-methyl-1h-indole-7-carboxaldehyde, 1h-indole-7-carboxaldehyde, 1-methyl-9ci
PubChem CID 11040979
IUPAC Name 1-methylindole-7-carbaldehyde
SMILES CN1C=CC2=C1C(=CC=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-7-carbaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.